missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXR1 Monoclonal Antibody (OTI5A5), TrueMAB™, OriGene
Description
FOXR1 Monoclonal antibody specifically detects FOXR1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | FOXR1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI5A5 |
| Concentration | 0.5 to 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | Foxr1 |
| Gene Accession No. | Q6PIV2 |
| Gene Alias | DLNB13; Forkhead box protein N5; forkhead box protein R1; forkhead box R1; forkhead box R1 variant 1; forkhead box R1 variant 2; Foxn5; FOXR1; Gm1115 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?