Learn More
FOXA2 Mouse anti-Human, Clone: OTI3C10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500073
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.Specifications
| FOXA2 | |
| Monoclonal | |
| Unconjugated | |
| FOXA2 | |
| HNF3B, TCF3B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3170 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9Y261 | |
| FOXA2 | |
| Recombit protein expressed in E.coli corresponding to amino acids 300-458 of human FOXA2 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.