Learn More
Fosfomycin Disodium Salt, TRC
Product Code. 30011296
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011296 10 g 10g
30007277 1 g 1g
17964255 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011296 Supplier Toronto Research Chemicals Supplier No. TRCF72750010G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26016-99-9
Molecular Formula C3 H5 O4 P . 2 Na
Molecular Weight (g/mol) 182.02
Synonym Fosfomycin sodium, Disodium (2R,3S)-(3-methyloxiran-2-yl)phosphonate
IUPAC Name disodium;[(2R,3S)-3-methyloxiran-2-yl]-dioxido-oxo-λ^{5}-phosphane
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P(=O)([O-])[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C3H7O4P.2Na/c1-2-3(7-2)8(4,5)6;;/h2-3H,1H3,(H2,4,5,6);;/q;2*+1/p-2/t2-,3+;;/m0../s1
Formula Weight 181.9721
Percent Purity >95%
Chemical Name or Material Fosfomycin Disodium Salt
Grade HPLC