Learn More
Fosfluconazole, MedChemExpress
Product Code. 16489666
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Product Code. 16489666
Brand: Medchem Express HY100666
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Fosfluconazole is a prodrug of Fluconazole that is widely used as an antifungal agent.
Fosfluconazole is a prodrug of Fluconazole that is widely used as an antifungal agent.
Specifications
Chemical Name or Material Fosfluconazole
Physical Form Powder
CAS 194798-83-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H13F2N6O4P
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 6.2 mg/mL (16.05 mM; Need ultrasonic and warming)
SMILES FC1=CC=C(C(CN2N=CN=C2)(OP(O)(O)=O)CN3N=CN=C3)C(F)=C1
Molecular Weight (g/mol) 386.25
Formula Weight 386.25
Percent Purity 98.0%
Grade Research