missing translation for 'onlineSavingsMsg'
Learn More

Fosfluconazole, MedChemExpress

Product Code. 16482626
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16482626

Brand: Medchem Express HY100666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fosfluconazole is a prodrug of Fluconazole that is widely used as an antifungal agent.

Fosfluconazole is a prodrug of Fluconazole that is widely used as an antifungal agent.

Specifications

Chemical Name or Material Fosfluconazole
Physical Form Powder
CAS 194798-83-9
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C13H13F2N6O4P
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 6.2 mg/mL (16.05 mM; Need ultrasonic and warming)
SMILES FC1=CC=C(C(CN2N=CN=C2)(OP(O)(O)=O)CN3N=CN=C3)C(F)=C1
Molecular Weight (g/mol) 386.25
Formula Weight 386.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.