Learn More
DiscoverX Forskolin
Description
Forskolin is a labdane diterpene produced by the Indian Coleus plant, Coleus forskohlii. It is commonly used to raise levels of cyclic AMP (cAMP) associated with the study of cell physiology mechanisms. It acts by activating the enzyme adenyl cyclase to increase intracellular levels of this second messenger, which is necessary for the proper biological response of cells to hormones and other extracellular signals. Provided as 2 vials of 0.25mg dry powder each.
Specifications
Specifications
| Quantity | 2 x 0.25 mg |