Learn More
DiscoverX Forskolin
Product Code. 17322349
Encompass
Change view
Click to view available options
Quantity:
2 x 0.25 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17322349 2 x 0.25 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17322349 Supplier DiscoverX Supplier No. 920005
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Forskolin is an activator of adenylyl cyclases. Provided as 2 vials of 0.25mg dry powder each.
Forskolin is a labdane diterpene produced by the Indian Coleus plant, Coleus forskohlii. It is commonly used to raise levels of cyclic AMP (cAMP) associated with the study of cell physiology mechanisms. It acts by activating the enzyme adenyl cyclase to increase intracellular levels of this second messenger, which is necessary for the proper biological response of cells to hormones and other extracellular signals. Provided as 2 vials of 0.25mg dry powder each.
Specifications
Quantity 2 x 0.25 mg