missing translation for 'onlineSavingsMsg'
Learn More

Formononetin, MedChemExpress

Product Code. p-200099366
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16471496

Brand: Medchem Express HYN0183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Formononetin is a potent FGFR2 inhibitor with an IC50 of ∼4.31 μM. Formononetin potently inhibits angiogenesis and tumor growth.

Formononetin is a potent FGFR2 inhibitor with an IC50 of ∼4.31 μM. Formononetin potently inhibits angiogenesis and tumor growth.

Specifications

Chemical Name or Material Formononetin
CAS 485-72-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H12O4
Quantity 200 mg
Synonym Biochanin B Flavosil Formononetol
Solubility Information DMSO : ≥ 35 mg/mL (130.47 mM)
SMILES O=C1C(C2=CC=C(OC)C=C2)=COC3=CC(O)=CC=C13
Molecular Weight (g/mol) 268.26
Formula Weight 268.26
Percent Purity 97.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.