missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Format Reticle MC

Product Code. 10654283
Click to view available options
Depth (Metric):
26 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10654283

Brand: Zeiss™ 4540750000000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for use with Zeiss Microscopes

Format Reticle MC
  • 2.5x
  • Depth: 26mm
  • Specifications

    Type Format Reticle MC
    Depth (Metric) 26 mm
    Magnification Power 2.5X
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.