Learn More
FOLR3 Mouse anti-Human, Clone: OTI7E10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811544S
Description
FOLR3 Monoclonal antibody specifically detects FOLR3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| FOLR3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41439 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2352 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7E10 | |
| Unconjugated | |
| FOLR3 | |
| Folate receptor 3; folate receptor 3 (gamma); folate receptor gamma; FOLR3; FR gamma; FR- gamma; FR-G; FR-gamma; gamma-hFR | |
| Full length human recombinant protein of human FOLR3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.