Learn More
Uvex Fold-flat Mask silv-Air p 8203 FFP2
Product Code. 17592955 Shop All Uvex Products
Change view
Click to view available options
Filter Type: FFP2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17592955 Filter Type: FFP2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17592955 Supplier Uvex Supplier No. 8708203
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering flat-fold mask
  • Hydrophobic material coating
  • Seamless headband for a comfortable, secure fit
  • Flexible, adjustable integrated nose clip
  • Foam layers in chin and nose area for optimally secure fit
  • Individually sealed
  • Color: silver
Protection Features
  • FFP2 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Seamless headband for a comfortable, secure fit
  • Flexible, adjustable integrated nose clip
  • Foam padding on the chin and nose for optimally secure fit
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Disposable Single-use
Material Polypropylene, Polyester
With Exhalation Valve No
Filter Efficiency FFP2
Sterility Non-sterile