Description
General Features
- Particle-filtering flat-fold mask
- Hydrophobic material coating
- Seamless headband for a comfortable, secure fit
- Flexible, adjustable integrated nose clip
- Foam layers in chin and nose area for optimally secure fit
- Individually sealed
- Color: silver
Protection Features
- FFP2 respirator certified in accordance with EN 149:2001+A1:2009
- D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
- Not reusable
Comfort Features
- Seamless headband for a comfortable, secure fit
- Flexible, adjustable integrated nose clip
- Foam padding on the chin and nose for optimally secure fit
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 149 (D marked) |
| Disposable | Single-use |
| Material | Polypropylene, Polyester |
| With Exhalation Valve | No |
| Filter Efficiency | FFP2 |
| Sterility | Non-sterile |