Learn More
Fodosine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Fodosine Hydrochloride |
| CAS | 284490-13-7 |
| Molecular Formula | C11H14N4O4 . HCl |
| Quantity | 1 mg |
| Synonym | (9CI)-7-[(2S,3S,4R,5R)-3,4-Dihydroxy-5-(hydroxymethyl)-2-pyrrolidinyl]-1,5-dihydro-, monohydrochloride 4H-pyrrolo[3,2-d]pyrimidin-4-one, BCX 1770, BCX 1777 |
| InChI Formula | InChI=1S/C11H14N4O4.ClH/c16-2-5-9(17)10(18)7(15-5)4-1-12-8-6(4)13-3-14-11(8)19;/h1,3,5,7,9-10,12,15-18H,2H2,(H,13,14,19);1H/t5-,7+,9-,10+;/m1./s1 |
| SMILES | Cl.OC[C@H]1N[C@H]([C@H](O)[C@@H]1O)c2c[nH]c3C(=O)NC=Nc23 |
| Molecular Weight (g/mol) | 302.71 |
| Formula Weight | 302.0782 |