missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FNDC8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18046571
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18046571 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18046571 Supplier Invitrogen™ Supplier No. PA5144882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EVAKTQENEL PEAKNRPWIF NKILGTTVKL MELKPNTCYC LSVRAANTAG VGKWCKPYKF ATLATDFSSF PENYPIQITV RRKEPRQKIV SIGPEEMRRL EDLEYLFPC Antigen sequence identity: Mouse 72%; Rat 72%.

FNDC8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNDC8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FNDC8
Gene Accession No. Q8TC99
Gene Alias 4930466G16Rik; fibronectin type III domain containing 8; fibronectin type III domain-containing protein 8; FNDC8; RGD1310708
Gene Symbols FNDC8
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 216-324. Recombinant protein control fragment (Product #RP-110145).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.