missing translation for 'onlineSavingsMsg'
Learn More

FN-1501, MedChemExpress

Product Code. p-200097059
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497147

Brand: Medchem Express HY111361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FN-1501 is a potent inhibitor of FLT3 and CDK, with IC50s of 2.47, 0.85, 1.96, and 0.28 nM for CDK2/cyclin A, CDK4/cyclin D1, CDK6/cyclin D1 and FLT3, respectively. FN-1501 has anticancer activity.

FN-1501 is a potent inhibitor of FLT3 and CDK, with IC50s of 2.47, 0.85, 1.96, and 0.28 nM for CDK2/cyclin A, CDK4/cyclin D1, CDK6/cyclin D1 and FLT3, respectively. FN-1501 has anticancer activity.

Specifications

Chemical Name or Material FN-1501
Physical Form Solid
CAS 1429515-59-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H25N9O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (115.88 mM)
SMILES O=C(C1=NNC=C1NC2=NC=NC3=C2C=CN3)NC4=CC=C(CN5CCN(C)CC5)C=C4
Molecular Weight (g/mol) 431.49
Formula Weight 431.49
Percent Purity 99.71%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.