Learn More
MedChemExpress Fmoc-N-Me-Phe-OH
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Purity | 97.0% |
| CAS | 77128-73-5 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 401.46 g/mol |
| Product Type | Reference compound |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |