missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fmoc-L-Tryptophan(Boc), 10/Package
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Format | Cartridge |
| Quantity | 10 Cartridge/Pack |
| Product Type | Amino Acid Cartridge |
| Content And Storage | Each package contains 10 cartridges. |
| For Use With (Equipment) | 433A Peptide Synthesizer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?