missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FMN2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16637014
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16637014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16637014 Supplier Invitrogen™ Supplier No. PA5100530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FMN2.

FMN2, a novel formin homology protein of the cappuccino subfamily, is highly conserved between evolutionarily diverse vertebrates. FMN2 bears a high degree of similarity to formin-1 which is the founding member of formin homology proteins. FMN2 plays a crucial role in maintenance of the meiotic spindle. The protein is widely expressed in human fetal brain, adult whole brain, hypothalamus, retina, pancreatic islet and germinal-center B cells. Pathological role of the protein is identified in various tumors like parathyloid tumor, glioblastoma, retinoblastoma and chondrosarcoma. FMN2 mutations are often a cause of unexplained infertility in women. The gene encoding for human FMN2 is localized in the chromosomal region 1q43.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FMN2
Gene Accession No. Q9JL04, Q9NZ56
Gene Alias AU024104; FMN2; formin 2; formin-2; RP11-90L13.1
Gene Symbols FMN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FMN2(Accession Q9NZ56), corresponding to amino acid residues M1-T35.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360457, 54418, 56776
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.