Learn More
FMA-Fluorescein Mercuric Acetate (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | FMA-Fluorescein Mercuric Acetate |
| CAS | 32382-27-7 |
| Molecular Formula | C24H16Hg2O9 |
| Quantity | 100 mg |
| Synonym | bis(acetato-κO)[μ-(3′,6′-dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-4′,5′-diyl)]di-Mercury, |
| InChI Formula | InChI=1S/C20H10O5.2C2H4O2.2Hg/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20;2*1-2(3)4;;/h1-8,21-22H;2*1H3,(H,3,4);;/q;;;2*+1/p-2 |
| SMILES | O=C1OC2(C3=CC=C(O)C([Hg]OC(=O)C)=C3OC4=C([Hg]OC(=O)C)C(O)=CC=C42)C=5C=CC=CC15 |
| Molecular Weight (g/mol) | 849.58 |