Learn More
Fluspirilene, MedChemExpress
Product Code. 16431128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431128 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16431128 Supplier Medchem Express Supplier No. HYB1655
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Fluspirilene is a non-competitive antagonist of L-type calcium channels with an IC50 of 0.03 μM. Fluspirileneis a long-acting injectable depot antipsychotic drug used for schizophrenia.
Fluspirilene is a non-competitive antagonist of L-type calcium channels with an IC50 of 0.03 μM. Fluspirileneis a long-acting injectable depot antipsychotic drug used for schizophrenia.
Specifications
Chemical Name or Material Fluspirilene
CAS 1841-19-6
Color White
Purity Grade Notes Research
Molecular Formula C29H31F2N3O
Quantity 5 mg
Synonym R 6218 Redeptin
SMILES O=C1NCN(C2=CC=CC=C2)C13CCN(CCCC(C4=CC=C(F)C=C4)C5=CC=C(F)C=C5)CC3
Molecular Weight (g/mol) 475.57
Formula Weight 475.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation