Learn More
Flurandrenolone Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Flurandrenolone acetate |
| CAS | 2802-11-1 |
| Molecular Formula | C26 H35 F O7 |
| Quantity | 1 mg |
| Synonym | (6α,11β,16α)-21-(acetyloxy)-6-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregn-4-ene-3,20-dione (9CI), Pregn-4-ene-3,20-dione, 6α-fluoro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-acetate (8CI), 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv., 6α-Fluoro-16α-hydroxyhydrocortisone 16,17-acetonide 21-acetate, Flurandrenolone acetate, 6α-Fluoro-11β-hydroxy-16α,17-(1-methylethylidenedioxy)-3,20-dioxopregn-4-en-21-yl Acetate |
| InChI Formula | InChI=1S/C26H35FO7/c1-13(28)32-12-20(31)26-21(33-23(2,3)34-26)10-16-15-9-18(27)17-8-14(29)6-7-24(17,4)22(15)19(30)11-25(16,26)5/h8,15-16,18-19,21-22,30H,6-7,9-12H2,1-5H3/t15-,16-,18-,19-,21+,22+,24-,25-,26+/m0/s1 |
| SMILES | CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)CC[C@]5(C)[C@H]4[C@@H](O)C[C@]23C |
| Molecular Weight (g/mol) | 478.55 |
| Formula Weight | 478.2367 |