missing translation for 'onlineSavingsMsg'
Learn More

Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%

Product Code. p-7046945
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383816 1 g 1g
11393816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 11383816 Supplier Thermo Scientific Alfa Aesar Supplier No. L16891.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 28523-86-6 | C4H3F7O | 200.056 g/mol

Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28523-86-6
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.056
MDL Number MFCD00153189
InChI Key DFEYYRMXOJXZRJ-UHFFFAOYSA-N
Synonym sevoflurane, sevofluran, ultane, 1,1,1,3,3,3-hexafluoro-2-fluoromethoxy propane, sevoflurano, sevofluranum, sevorane, sojourn, mr6s4, sevofluranum inn-latin
PubChem CID 5206
ChEBI CHEBI:9130
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
SMILES C(OC(C(F)(F)F)C(F)(F)F)F

Specifications

Density 1.505
Boiling Point 58°C
Quantity 1 g
Beilstein 2041023
Merck Index 14,8475
Solubility Information Slightly miscible with water.
Formula Weight 200.06
Percent Purity ≥98%
Chemical Name or Material Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.