missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fluorescein-5-maleimide

Product Code. 16464667
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16464667

Brand: MedChemExpress HYD0098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fluorescein-5-maleimide is a fluorescent thiol-reactive dye used to conjugate fluorescein to proteins (excitation: 494 nm, emission: 519 nm).

Specifications

Quantity 50 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 427.36 g/mol
Solubility DMSO : 50 mg/mL (117.00 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 75350-46-8
Synonym N-(5-Fluoresceinyl)maleimide
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.