Learn More
Fluorene-1-carbamic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Fluorene-1-carbamic Acid Ethyl Ester |
| CAS | 28314-06-9 |
| Molecular Formula | C16 H15 N O2 |
| Quantity | 50 mg |
| Synonym | Ethyl 9H-Fluoren-1-ylcarbamate |
| InChI Formula | InChI=1S/C16H15NO2/c1-2-19-16(18)17-15-9-5-8-13-12-7-4-3-6-11(12)10-14(13)15/h3-9H,2,10H2,1H3,(H,17,18) |
| SMILES | CCOC(=O)Nc1cccc2c1Cc3ccccc23 |
| IUPAC Name | ethyl N-(9H-fluoren-1-yl)carbamate |
| Molecular Weight (g/mol) | 253.2958 |
| Formula Weight | 253.1103 |