Learn More
Fluocortolone, TRC
Specifications
Specifications
| Chemical Name or Material | Fluocortolone |
| CAS | 152-97-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H29 F O4 |
| Quantity | 5 mg |
| Synonym | Pregna-1,4-diene-3,20-dione, 6-fluoro-11,21-dihydroxy-16-methyl-, (6α,11β,16α)-, Pregna-1,4-diene-3,20-dione, 6α-fluoro-11β,21-dihydroxy-16α-methyl- (7CI,8CI), 16α-Methyl-6α-fluoro-Δ1,4-pregnadien-11β,21-diol-3,20-dione, 6α-Fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, 6α-Fluoro-16α-methyl-1,4-pregnadiene-11β,21-diol-3,20-dione, 6α-Fluoro-16α-methyl-Δ1,4-pregnadiene-11β,21-diol-3,20-dione, 6α-Fluoro-16α-methyl-Δ1-corticosterone, Flucortolone, Fluocortolon, Fluocortolone, SH 742, Ultralan oral |
| InChI Formula | InChI=1S/C22H29FO4/c1-11-6-14-13-8-16(23)15-7-12(25)4-5-21(15,2)20(13)17(26)9-22(14,3)19(11)18(27)10-24/h4-5,7,11,13-14,16-17,19-20,24,26H,6,8-10H2,1-3H3/t11-,13+,14+,16+,17+,19-,20-,21+,22+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]2(C)[C@H]1C(=O)CO |
| IUPAC Name | (6S,8S,9S,10R,11S,13S,14S,16R,17S)-6-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 376.46 |
| Show More |