Learn More
MedChemExpress Fluo-4 AM
Specifications
Specifications
| Content And Storage | -20°C, sealed storage, away from moisture and light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Product Type | Dye Reagents |
| Quantity | 100 μg |
| Shipping Condition | Room temperature |
| Molecular Weight (g/mol) | 1096.94 g/mol |
| CAS | 273221-67-3 |