Learn More
Flunisolide, MedChemExpress
Product Code. 16438626
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16438626
Brand: Medchem Express HYB1121
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Flunisolide is a corticosteroid, which is an orally active glucocorticoid receptor activator with anti-inflammatory activity. Flunisolide can induce eosinophil apoptosis, and is used for the research of asthma or rhinitis, and inflammation.
Flunisolide is a corticosteroid, which is an orally active glucocorticoid receptor activator with anti-inflammatory activity. Flunisolide can induce eosinophil apoptosis, and is used for the research of asthma or rhinitis, and inflammation.
Specifications
Chemical Name or Material Flunisolide
CAS 3385-03-3
Color White
Purity Grade Notes Research
Molecular Formula C24H31FO6
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (287.69 mM; Need ultrasonic)
SMILES C[C@@]12[C@@]3(C(CO)=O)[C@@](OC(C)(O3)C)([H])C[C@@]1([H])[C@]4([H])C[C@H](F)C5=CC(C=C[C@]5(C)[C@@]4([H])[C@@H](O)C2)=O
Molecular Weight (g/mol) 434.5
Formula Weight 434.5
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation