missing translation for 'onlineSavingsMsg'
Learn More

Flufenamic Acid, TRC

Product Code. 30006407
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006407 5 g 5g
30008548 50 mg 50mg
30004971 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006407 Supplier Toronto Research Chemicals Supplier No. TRCF4350005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-78-9
Formula Weight 281.0664
InChI Formula InChI=1S/C14H10F3NO2/c15-14(16,17)9-4-3-5-10(8-9)18-12-7-2-1-6-11(12)13(19)20/h1-8,18H,(H,19,20)
IUPAC Name 2-[3-(trifluoromethyl)anilino]benzoic acid
Molecular Formula C14 H10 F3 N O2
Molecular Weight (g/mol) 281.23
Percent Purity >95%
SMILES OC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F
Synonym Flufenamic Acid, Etofenamate Imp. A (EP), 2-[3-(Trifluoromethyl)anilino]benzoic acid, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid, 3'-Trifluoromethyl-N-phenylanthranilic acid, 3'-Trifluoromethyldiphenylamine-2-carboxylic acid, ANT-1, Achless, Ansatin, Arlef, CI 440, CN 27544, Flufenamic acid, Fluphenamic acid, Fullsafe, INF 1837, Meralen, Movilizin, N-(α,α,α-Trifluoro-m-tolyl)anthranilic acid, N-(m-Trifluoromethylphenyl)-2-aminobenzoic acid, N-[3-(Trifluoromethyl)phenyl]anthranilic acid, NSC 219007, NSC 82699, Paraflu, Parlef, Parlif, Pinox, Plostene, Ristogen, Sastridex, Surika, Tecramine
Chemical Name or Material Flufenamic Acid
Grade HPLC
Show More Show Less

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.