missing translation for 'onlineSavingsMsg'
Learn More

Fludazonium chloride, MedChemExpress

Product Code. p-200100844
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470736 20 mg 20mg
16443209 1 mg 1mg
16422428 5 mg 5mg
16400229 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470736 Supplier Medchem Express Supplier No. HYU00181

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Fludazonium chloride (R23633) is an anti-fungal agent, which can be used in the treatment and prevention of superficial and systemic fungal infections.

Fludazonium chloride (R23633) is an anti-fungal agent, which can be used in the treatment and prevention of superficial and systemic fungal infections.

Specifications

Chemical Name or Material Fludazonium chloride
Physical Form Solid
CAS 53597-28-7
Color White
Quantity 20 mg
Purity Grade Notes Research
Molecular Formula C26H20Cl5FN2O2
For Use With (Application) COVID-19-immunoregulation
Synonym R23633
SMILES O=C(C1=CC=C(F)C=C1)C[N+]2=CN(CC(C3=CC=C(Cl)C=C3Cl)OCC4=CC=C(Cl)C=C4Cl)C=C2.[Cl-]
Molecular Weight (g/mol) 588.71
Formula Weight 588.71
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.