Learn More
Fludazonium chloride, MedChemExpress
Product Code. 16470736
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470736 20 mg 20mg
16443209 1 mg 1mg
16422428 5 mg 5mg
16400229 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16470736 Lieferant Medchem Express Lieferanten-Nr. HYU00181
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Fludazonium chloride (R23633) is an anti-fungal agent, which can be used in the treatment and prevention of superficial and systemic fungal infections.
Fludazonium chloride (R23633) is an anti-fungal agent, which can be used in the treatment and prevention of superficial and systemic fungal infections.
Spezifikation
Chemical Name or Material Fludazonium chloride
Physical Form Solid
CAS 53597-28-7
Color White
Quantity 20 mg
Purity Grade Notes Research
Molecular Formula C26H20Cl5FN2O2
For Use With (Application) COVID-19-immunoregulation
Synonym R23633
SMILES O=C(C1=CC=C(F)C=C1)C[N+]2=CN(CC(C3=CC=C(Cl)C=C3Cl)OCC4=CC=C(Cl)C=C4Cl)C=C2.[Cl-]
Molecular Weight (g/mol) 588.71
Formula Weight 588.71
Percent Purity 97.0%
Grade Research