Learn More
R&D Systems™ FlowX™ Mouse Regulatory T Cell Multi-Color Flow Kit
Use for the single-step staining of mouse regulatory T cells (1-9)
Brand:  R&D Systems™ FMC022
Additional Details : Weight : 0.09700kg
Product Code. 15703204
  • 735.00 € / 25 tests
Estimated Shipment: 24-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Contains conjugated antibodies to CD25-APC (Clone 280406), FoxP3-PE (Clone 1054C), CD4-CFS (Clone GK1.5)
Specifications
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date.
25 Tests