Learn More
R&D Systems™ FlowX Human Th2 Cell Multi-Color Flow Cytometry Kit
R&D Systems™ FlowX Human Th2 Cell Multi-Color Flow Cytometry Kit is designed for the flow cytometric analysis of Th2 cells using five fluorochrome-conjugated antibodies.
Brand: R&D Systems™ FMC011B
Additional Details : Weight : 0.00970kg
Description
- This kit contains five conjugated antibodies and buffers that can be used for staining of human Th2 cells (1-6)
- Positive Markers:
- 125μL of CD4 Alexa Fluor™ 700 Mouse IgG2A (Clone 11830)
- 125μL of IL-10 Alexa Fluor™ 488 Rabbit IgG (Clone 2050B)
- 250μL of CCR3-PE Rat IgG2A (Clone 61828)
- 125μL of GATA-3 Alexa Fluor™ 594 Mouse IgG2B (Clone 634919)
- 125μL of Phospho-STAT6 Alexa Fluor™ 647 Rabbit IgG (Clone 1248D)
- Fixation/Permeabilization Buffer (1X):
- 30mL of 1X fixation buffer.
- Staining Buffer (1X):
- 50mL of a 1X staining buffer
- Some components in this kit contain formaldehyde which is a suspected carcinogen.
- Avoid contact with skin, eyes, and mucous membranes, and avoid inhaling fumes.
- In case of contact, wash immediately with water and seek medical advice.
- Some components in this kit contain sodium azide which may react with lead and copper plumbing to form explosive metallic azides.
- Flush with large volumes of water during disposal.
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. | |
25 |