Learn More
R&D Systems™ FlowX Human Th1 Cell Multi-Color Flow Cytometry Kit
R&D Systems™ FlowX Human Th1 Cell Multi-Color Flow Cytometry Kit is designed for the flow cytometric analysis of Th1 cells using five fluorochrome-conjugated antibodies.
Brand: R&D Systems™ FMC009B
Additional Details : Weight : 0.09700kg
Description
- This kit contains five conjugated antibodies and buffers that can be used for staining of human Th1 cells (1-5)
- Positive Markers:
- 125μL of CD4 Alexa Fluor™ 700 Mouse IgG2A (Clone 11830)
- 125μL of TIM-3 Alexa Fluor™ 594 Rat IgG2A (Clone 344823)
- 250μL of IL-12 Rβ2-PE Mouse IgG1 (Clone 305719)
- 250μL of IFN-γ-PerCP Mouse IgG2B (Clone 25723)
- 125μL of T-bet Alexa Fluor™ 488 Mouse IgG1 (Clone 525831)
- FoxP3/Transcription Factor Fixation Concentrate (4X):
- 8mL of a formaldehyde solution.
- FoxP3/Transcription Factor Fixation Diluent:
- 25mL of a buffered detergent
- FoxP3/Transcription Factor Permeabilization and Wash Buffer (10X)
- 25mL of a buffered protein base with detergent and preservatives. May contain a precipitate but will not affect product performance.
- Staining Buffer (1X):
- 50mL of a 1X staining buffer
- Some components in this kit contain formaldehyde which is a suspected carcinogen.
- Avoid contact with skin, eyes, and mucous membranes, and avoid inhaling fumes.
- In case of contact, wash immediately with water and seek medical advice.
- Some components in this kit contain sodium azide which may react with lead and copper plumbing to form explosive metallic azides.
- Flush with large volumes of water during disposal.
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. | |
25 |