Learn More
Cole-Parmer™ Stuart™ Florentine Flask For Stuart™ Rotary Evaporators
Description
- For use with Stuart™ rotary evaporators
- Flat bottom
- 29/32 socket
Specifications
Specifications
| Description | Evaporating flask |
| Capacity (Metric) | 1 L |
| Material | Glass |
| Standard Taper Joint | 29/32 |
| Bottom Shape | Flat |