Learn More
STEMCELL Technologies Single Flow Meter
Product Code. 16231962
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
For Use With (Application):
Cell Culture
missing translation for 'unitSize'
Each
Product Code. 16231962
missing translation for 'mfr': STEMCELL Technologies 27311
Please to purchase this item. Need a web account? Register with us today!
Meter to control gas flow into hypoxia incubator chamber
  • Single Flow Meter allows for precise control of gas flow to generate a hypoxic environment for tissue culture using the Hypoxia Incubator Chamber (Catalog #27310)
  • Features of the Single Flow Meter include:
  • Same precision as the flow meter used for full size continuous flow carbon dioxide incubator chambers
  • Calibrated for 5 - 50 liters per minute (LPM)
Specifications
For Use With (Application) Cell Culture
Length (Metric) 1 m