Learn More
Flibanserin-d4 HCl (ethylene-d4), CDN
Specifications
Specifications
| Chemical Name or Material | Flibanserin-D4 (ethylene-D4) Hydrochloride |
| CAS | 2748630-46-6 |
| Molecular Formula | C20 D4 H17 F3 N4 O . Cl H |
| Quantity | 0.01 g |
| Synonym | 1,3-Dihydro-1-[2-[4-[3-(trifluoromethyl)phenyl]-1-piperazinyl]ethyl-1,1,2,2-d4]-2 H-benzimidazol-2-one hydrochloride (1:1), Flibanserin-d4 hydrochloride, Flibanserin-D4 (ethylene-D4) hydrochloride, Flibanserin-d4 HCl (ethylene-d4) |
| InChI Formula | InChI=1 S/C20H21F3N4O.ClH/c21-20(22,23)15-4-3-5-16(14-15)26-11-8-25(9-12-26)10-13-27-18-7-2-1-6-17(18)24-19(27)28;/h1-7,14 H,8-13H2,(H,24,28);1 H/i10D2,13D2; |
| SMILES | Cl.[2 H]C([2 H])(N1CCN(CC1)c2cccc(c2)C(F)(F)F)C([2 H])([2 H])N3C(=O)Nc4ccccc34 |
| IUPAC Name | 3-[1,1,2,2-tetradeuterio-2-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]ethyl]-1 H-benzimidazol-2-one;hydrochloride |
| Molecular Weight (g/mol) | 430.8877 |
| Formula Weight | 430.1685 g/mol |
| Show More |