Learn More
FLI-06, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4,5,6,7,8-Hexahydro-2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-3-quinolinecarboxylic Acid Cyclohexyl Ester |
| CAS | 313967-18-9 |
| Molecular Formula | C25H30N2O5 |
| Quantity | 25 mg |
| Synonym | FLI-06, 1,4,5,6,7,8-Hexahydro-2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-3-quinolinecarboxylic Acid Cyclohexyl Ester |
| InChI Formula | InChI=1S/C25H30N2O5/c1-15-21(24(29)32-18-7-5-4-6-8-18)22(16-9-11-17(12-10-16)27(30)31)23-19(26-15)13-25(2,3)14-20(23)28/h9-12,18,22,26H,4-8,13-14H2,1-3H3 |
| SMILES | CC1=C(C(C2=C(CC(C)(C)CC2=O)N1)c3ccc(cc3)[N+](=O)[O-])C(=O)OC4CCCCC4 |
| IUPAC Name | cyclohexyl 2,7,7-trimethyl-4-(4-nitrophenyl)-5-oxo-1,4,6,8-tetrahydroquinoline-3-carboxylate |
| Molecular Weight (g/mol) | 438.52 |
| Formula Weight | 438.2155 |