Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF30plus
Flexible temperature sensor for positioning in chamber or in load.
Brand: Memmert™ UF30plusH8
Additional Details : Weight : 0.01200kg
Description
- Positioned flexibly in chamber or load
- For local temperature measurement (up to 1 additional sensor is possible)
- The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
- For Memmert Ovens
Specifications
UF30plus Oven | |
Temperature Sensor |