Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF30plus
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UF30plusH8
Additional Details : Weight : 0.01200kg
Product Code. 17519305
  • 212.00 € / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UF30plus Oven
Temperature Sensor