Learn More
Heidolph™ Flask PolyBlock
Product Code. 11342723 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
1 x 250 mL Flask
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11342723 1 x 250 mL Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11342723 Supplier Heidolph™ Supplier No. 5058002000
  • ONLINE EXCLUSIVE PRICE
    273.00 €
    254.00 € / Pack of 1
    Save 19.00 €
    6% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Accommodate specifically sized vessels for your StarFish magnetic stirrer workstation. Heidolph™ Flask PolyBlock, is one of up to 5 StarFish PolyBlock segments that can be mixed at a single stirrer workstation to create various arrays of vessel size and type.
  • Accommodates one specific flask size.
  • One of up to 5 segments per StarFish array.
TRUSTED_SUSTAINABILITY
Specifications
Description PolyBlock
For Use With (Equipment) Starfish Workstation, MR-Hei Magnetic Stirrers
Holds 1 x 250 mL Flask