Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Silica
Product Code. 16390146 Shop All Buchi Products
Change view
Click to view available options
Includes:
1 Pc.
10 Pc.
12 Pc.
18 Pc.
2 Pc.
20 Pc.
3 Pc.
6 Pc.
Bed Weight (Metric):
4 g
12 g
25 g
120 g
220 g
800 g
1600 g
3000 g
Unit Size:
Each
Pack of 10
Pack of 12
Pack of 18
Pack of 2
Pack of 20
Pack of 3
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Bed Weight (Metric) Unit Size
16390146 1 Pc. 3000 g Each
15857051 10 Pc. 120 g Pack of 10
16330146 18 Pc. 12 g Pack of 18
16380146 2 Pc. 1600 g Pack of 2
15867051 6 Pc. 220 g Pack of 6
16340146 12 Pc. 25 g Pack of 12
16320146 20 Pc. 4 g Pack of 20
16370146 3 Pc. 800 g Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16390146 Supplier BUCHI Supplier No. 140000031
  • ONLINE EXCLUSIVE PRICE
    687.00 €
    651.91 € / Pack of 1
    Save 35.09 €
    5% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Type Silica
Includes 1 Pc.
Resolution 40 to 63 μm
Bed Weight (Metric) 3000 g
Column Type Silica
For Use With (Application) Basic Applications, Polar Compounds
For Use With (Equipment) Any Flash System