Learn More
Fisherbrand™ Fixed Tube Rack
Product Code. 15793798 Shop All Fisherbrand Products
Click to view available options
Holds:
21 × 30mm dia., 50mL tubes, Tubes (5 mL)
27 x 25 mL Tubes, Tubes (25 mL)
40 x 15 mL Tubes
Unit Size:
Each
Product Code. 15793798
Please to purchase this item. Need a web account? Register with us today!
For use with Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
Maximum 2 per platform
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixed Tube Rack
Holds 27 x 25 mL Tubes, Tubes (25 mL)
Positions 27
For Use With (Equipment) Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
Dimensions (L x W x H) 262 x 112 x 140 mm
Diameter (Metric) Outer 22 mm