Learn More
Fisherbrand™ Fixed Tube Rack
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Holds:
21 × 30mm dia., 50mL tubes, Tubes (5 mL)
27 x 25 mL Tubes, Tubes (25 mL)
40 x 15 mL Tubes
missing translation for 'unitSize'
Each
Description
Maximum 2 per platform
Specifications
Specifications
| Product Type | Fixed Tube Rack |
| Holds | 21 × 30mm dia., 50mL tubes, Tubes (5 mL) |
| Positions | 21 |
| For Use With (Equipment) | Fisherbrand Multi-Platform Shaker and Microplate Mini Shaker |
| Dimensions (L x W x H) | 262 x 112 x 140 mm |
| Diameter (Metric) Outer | 30 mm |