Learn More
Hirschmann™ Fitting Wrench for Valves
Product Code. 10287790
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10287790 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10287790 Supplier Hirschmann™ Supplier No. 9316901
Only null left
Add to basket
Add to basket
Valve tool with two sizes, suitable for any check valve in Hirschmann™ liquid handling devices.
  • For valve mounting on any Hirschmann™ instrument bottle top
Specifications
Type Valve Wrench
Includes 1