missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FITC Monoclonal antibody specifically detects FITC in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In-situ Hybridization
Specifications
Specifications
| Antigen | FITC |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM395 |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Fluorescein Isothiocyanate |
| Host Species | Mouse |
| Immunogen | Fluorescein isothiocyanate (FITC) conjugated to KLH |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?