Learn More
Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes
Product Code. 10579261
Change view
Click to view available options
Path Length:
1 mm
10 mm
Material:
Optical Glass
Quartz Suprasil 300
Suprasil Quartz
Quantity:
2/Pk.
Ea.
Unit Size:
Each
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Path Length Material Quantity Unit Size
10579261 10 mm Optical Glass 2/Pk. Each
10258560 1 mm Quartz Suprasil 300 Ea. Each
10340721 10 mm Quartz Suprasil 300 Ea. Each
11781784 10 mm Suprasil Quartz Ea. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10579261 Supplier Hellma™ Supplier No. 1508736
  • ONLINE EXCLUSIVE PRICE
    40.51 €
    34.97 € / Pack of 1
    Save 5.54 €
    13% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Includes
Fitted PTFE covers or tapered PTFE stoppers, except 14-385-906A and -902A, which come with glass cover plates
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
  • Quartz - high UV transmission and low thermal expansion
  • Glass - used in the visible range of the spectrum
  • Low values of refractive index and dispertion
  • Good resistance to chemicals
Specifications
Material Optical Glass
Quantity 2/Pk.
Path Length 10 mm