missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Seamless Knit Cotton/Polyester Gloves with Double-Sided PVC Dot Grip

Product Code. 10066401 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Large
Unit Size:
12 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10066401 Large 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10066401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in warehousing and package handling, general assembly, light duty general work situations, construction, and bottle handling.

  • Seamless knit shell is comfortable and flexible
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Double-sided PVC dots enhance grip and provide additional resistance to cuts and abrasions
  • Small size fits ladies well

Specifications

Construction Seamless Knit
Coating Material PVC Dots (2 Sides)
Color Natural, Blue
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Light Duty Tasks, Bottle Handling, Construction, General Assembly, and Warehousing
Glove Cuff Color Natural
Glove Cuff Style Knit
Series 37-C110DD
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.