Learn More
Fisherbrand™ Disposable High Tack Shoe Covers
High tack, disposable shoe covers provide superior protection
74.00 € - 77.70 €
Specifications
Material | Polyurethane |
---|---|
Latex Free | Yes |
Color | Blue |
Disposable | Single-use |
Sole Type | High Tack |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15421952
|
Fisherbrand™
|
Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15431952
|
Fisherbrand™
|
X-Large |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
High tack, disposable shoe covers provide superior protection
- Single-use
- Polyurethane
- Blue
- Latex-free
- High tack sole
Specifications
Polyurethane | |
Blue | |
High Tack |
Yes | |
Single-use |