Learn More
Fisherbrand™ Filter Cards for Shandon™ Cytospin™
Product Code. 15995742 Shop All Fisherbrand Products
Change view
Click to view available options
Filter Cards for Shandon Cytospin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15995742 Filter Cards for Shandon Cytospin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15995742
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
White cards are made from a blend of 100% cotton and cellulose fibers
  • Excellent absorbent padding for the collection of excess reagents or wash solution
  • Cellulose fibers improve the card's strength and durability during centrifugation and facilitate the removal of the filter card from the slide
Specifications
For Use With (Equipment) Shandon Cytospin
Type Filter Card