Learn More
Fisherbrand™ Gel Casting Pouches for FB-GC16-1 Vertical Gel Casters
Product Code. 11962975
Click to view available options
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
Product Code. 11962975
Brand: Fisherbrand™ GP2100
Please to purchase this item. Need a web account? Register with us today!
Maintain the quality of your current gel casting system with Fisherbrand™ Owl™ JGC-2 Gel Casting System Replacement Parts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Gel Casting
For Use With (Equipment) FB-GC16-1 Vertical Gel Casters
Type Gel Casting Pouch
Gel Dimensions 16 x 14 cm
Quantity 100/Pk.
Dimensions (L x W) 6.3 x 5.5 in. (16 x 14 cm)
Length (Metric) 16 cm
Width (English) 5.5 in.
Width (Metric) 14 cm