Learn More
ASECOS™ Q-Classic-30 Safety Storage Cabinet
Safe and approved cabinet for storage of flammable hazardous substances in working areas. Asecos™ Q-Classic-30 Safety Storage Cabinet is robust, and easy to handle, transport, and align.
Brand:  ASECOS™ 3100206130078
Product Code. 15211045
  • POA / Pack of 1
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Robust and long-lasting
    • Triple-hinged door
    • Scratch- and impact-resistant
  • Easy handling
    • Doors stop in any position
    • Self-closing in the event of fire
  • Locking doors
  • Easy transport
  • Easy alignment
  • Ventilation
Specifications
Specifications
FM Approved, 30 EN 14470-1, EKS / AK4 / EN 14727, CE
616 mm
Storage of hazardous (flammable) substances in the workplace
1196 mm
Fire safety storage cabinet
3 Tray Shelves and Bottom Tray, 4 Full Extension Drawers, 6 Full Extension Drawers
616 x 1196 x 1968 mm
1968 mm
Light Gray
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products