missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FIPI

Description
FIPI is a potent phosholipase D (PLD) inhibitor (IC50 values are 20 and 25 nM for PLD2 and PLD1 respectively). Ameliorates PLD2-driven suppression of membrane ruffling and cell spreading. Orally available.
Specifications
Specifications
| CAS | 1781834-93-2 |
| Quantity | 10 mg |
| Product Type | FIPI |
| Purity | >98% |
| Molecular Weight (g/mol) | 457.93 |
| Inhibitors | FIPI |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?