missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ratiolab™ Finntip 1000-5000 μL Pipette Tips
Click to view available options
Volume (Metric):
1000 to 5000 μL
Unit Size:
Pack of 1000
Description
- Uniform high quality and precision makes these tips fully effective
- Can be used with specially developed Multirack, Refill Systems, and Multiboxes
- Provides for simple and safe pipetting
- Can be used in the most sensitive applications
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to the controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Provided in packs of 1000
Specifications
Specifications
| Volume (Metric) | 1000 to 5000 μL |
| Color | Transparent |
| Autoclavable | Autoclavable |
| Format | Bulk |
| Quantity | 1000/pk |
| Product Type | Pipette tip |
| For Use With (Equipment) | Universal Fit, Both Manual or Electronic, Universal Fit |
| Product Line | FinnTip |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction